Learn More
MRGPRX2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MRGPRX2.
Manufacturer:  Abnova Corporation PAB16406
Catalog No. 89-118-278
Add to cart
Description
Description
Specifications
Specifications
MRGPRX2
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (20 ug/mL) The optimal working dilution should be determined by the end user.
MRGPRX2
MRGX2
Rabbit
Immunoaffinity purification
RUO
Primary
117194
Human
Immunohistochemistry (PFA fixed)
Rabbit polyclonal antibody raised against synthetic peptide of MRGPRX2.
In PBS (0.09% sodium azide)
Liquid
MRGPRX2
A synthetic peptide (conjugated with KLH) corresponding to human MRGPRX2.
50 ug
Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
C-terminal Cytoplasmic domain of human.