Learn More
MRPL20 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MRPL20.
Manufacturer: Abnova Corporation PAB17561
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. A pseudogene corresponding to this gene is found on chromosome 21q. [provided by RefSeq
Specifications
MRPL20 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of MRPL20. | |
In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
L20mt/MGC4779/MGC74465/MRP-L20 | |
Rabbit | |
100 μg | |
Primary | |
This antibody detects endogenous levels of total MRPL20 protein. | |
Liquid |
ELISA, Western Blot | |
Unconjugated | |
Western Blot (1:500-1:1000) ELISA (1:40000) The optimal working dilution should be determined by the end user. | |
MRPL20 | |
MRPL20 | |
A synthetic peptide corresponding to C-terminus of human MRPL20. | |
RUO | |
55052 | |
Human, Mouse, Rat |
Safety and Handling
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only