Learn More
MRPS24 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant MRPS24
Supplier: Abnova Corporation PAB28022
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein. A pseudogene corresponding to this gene is found on chromosome 11. [provided by RefSeq
Sequence: GTFPGCLADQLVLKRRGNQLEICAVVLRQLSPHKYYFLVGYSETLLSYFYKCPVRLHLQTVPSKVVYKYSpecifications
MRPS24 | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant MRPS24 | |
In PBS, pH 7.2, (40% glycerol, 0.02% sodium azide) | |
HSPC335/MRP-S24 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
64951 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunohistochemistry (1:20 - 1:50) Western Blot (1:100 - 1:250) The optimal working dilution should be determined by the end user. | |
MRPS24 | |
MRPS24 | |
Recombinant protein corresponding to amino acids of human MRPS24 | |
100 μL | |
Primary | |
Human | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only