Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSRA, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against full-length recombinant MSRA.

Manufacturer:  Abnova Corporation PAB1201

Catalog No. 89-115-520


Add to cart

Description

Description

This protein is ubiquitous and highly conserved. It carries out the enzymatic reduction of methionine sulfoxide to methionine. Human and animal studies have shown the highest levels of expression in kidney and nervous tissue. Its proposed function is the repair of oxidative damage to proteins to restore biological activity. Three transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Specifications

Specifications

MSRA
Unconjugated
Western Blot (1:1800) The optimal working dilution should be determined by the end user.
MSRA
MSRA
Recombinant protein corresponding to full length human MSRA.
RUO
Primary
4482
Western Blot
Rabbit polyclonal antibody raised against full length recombinant MSRA.
In HEPES, 150mM NaCl (50% glycerol, 0.01% BSA, 0.03% sodium azide)
Liquid
Rabbit
100 uL
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only