Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Myc Epitope Tag, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243691

Catalog No. NBP243691


Add to cart

Description

Description

Specifically detects Myc Epitope Tag in Human, Other samples, and it is validated for Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blotting

Myc Epitope Tag Polyclonal antibody specifically detects Myc Epitope Tag in Human, Non-species specific samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications

Specifications

Myc Epitope Tag
Unconjugated
PBS (pH 7.0) and 20% Glycerol with 0.01% Thimerosal
Rabbit
IgG
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Western Blot 1:5000-1:20000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:10000
Ion Exchange Chromatography
Synthetic peptide corresponding to a region within amino acids 410 and 419 of human c-Myc
Immunogen affinity purified
RUO
Primary
Human, Non-species specific
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only