Learn More
Myeloperoxidase, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against Myeloperoxidase.
Manufacturer:  Abnova Corporation PAB14010
Catalog No. 89-137-798
Add to cart
Description
Description
Specifications
Specifications
Myeloperoxidase
Unconjugated
Western Blot (1:4000) The optimal working dilution should be determined by the end user.
Liquid
Native from human neutrophil myeloperoxidase.
RUO
Primary
Neutrophil Myeloperoxidase.
Immunofluorescence
Rabbit polyclonal antibody raised against Myeloperoxidase.
In 10mM PBS, pH 7.2 (0.09% sodium azide)
Rabbit
1 mL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human