Learn More
Mylpf, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Mylpf.
Manufacturer:  Abnova Corporation PAB14878
Catalog No. 89-117-323
Add to cart
Description
Description
Specifications
Specifications
Mylpf
Unconjugated
Immunohistochemistry (5-10 ug/mL) The optimal working dilution should be determined by the end user.
Mylpf
2410014J02Rik|MLC-2|Mlc2
Rabbit
25 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Mouse
Immunohistochemistry (PFA fixed)
Rabbit polyclonal antibody raised against synthetic peptide of Mylpf.
In PBS (0.1% proclin, 2% Block Ace)
Liquid
Mylpf
A synthetic peptide corresponding to mouse Mylpf.
RUO
Primary
17907