Learn More
NANOS2 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NANOS2.
Manufacturer:  Abnova Corporation PAB13383
Catalog No. 89-116-630
Add to cart
Description
Description
Specifications
Specifications
NANOS2
Unconjugated
Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
NANOS2
NOS2
Rabbit
100 ug
Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human, Murine, Rat
Immunohistochemistry, Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of NANOS2.
In PBS (0.02% sodium azide)
Liquid
NANOS2
A synthetic peptide corresponding to N-terminus 13 amino acids of human NANOS2.
RUO
Primary
339345