Learn More
NANP Rabbit anti-Dog, Hamster, Human, Mouse, Rabbit, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NANP.
Manufacturer:  Abnova Corporation PAB16305
Catalog No. 89-118-179
Add to cart
Description
Description
Specifications
Specifications
NANP
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10-15 ug/mL) The optimal working dilution should be determined by the end user.
NANP
C20orf147|HDHD4|MGC26833|dJ694B14.3
Rabbit
Immunoaffinity purification
RUO
Primary
140838
Canine, Hamster, Human, Murine, Rabbit, Rat
Immunohistochemistry (PFA fixed)
Rabbit polyclonal antibody raised against synthetic peptide of NANP.
In PBS (0.09% sodium azide)
Liquid
NANP
A synthetic peptide (conjugated with KLH) corresponding to human NANP.
50 ug
Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Internal domain of human.