Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-NAP1L4, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236542

Catalog No. NBP236542


Add to cart

Description

Description

NAP1L4 Polyclonal antibody specifically detects NAP1L4 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).
Specifications

Specifications

NAP1L4
1.0 mg/mL
Immunoprecipitation 2 - 10 ug/mg, Western Blot (Negative)
NP_005960.1
hNAP2, MGC4565, NAP-2, NAP2L, NAP2NAP1L4b, nucleosome assembly protein 1-like 4, nucleosome assembly protein 1-like 4b, Nucleosome assembly protein 2
Rabbit
IgG
100 ul
Store at 4C. Do not freeze.
Polyclonal
Human
Immunoprecipitation, Western Blot
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
NAP1L4
The immunogen recognized by the antibody maps to a region between residue 125 to 175 of human Nucleosome Assembly Protein 2 using the numbering given in entry NP_005960.1.
Immunogen affinity purified
RUO
Primary
4676
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit