Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCK1/NCK2, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against recombinant NCK1/NCK2.

Manufacturer:  Abnova Corporation PAB12800

Catalog No. 89-116-098


Add to cart

Description

Description

Specifications

Specifications

NCK1/NCK2
1 mg/mL
Rabbit polyclonal antibody raised against recombinant NCK1/NCK2.
In HEPES, 150mM NaCl (50% glycerol, 0.01% BSA, 0.03% sodium azide)
MGC12668|NCK|NCKalpha
Rabbit
Affinity purification
RUO
Primary
4690|8440
Western Blot
Unconjugated
Western Blot (1:2000) The optimal working dilution should be determined by the end user.
Liquid
NCK1
Recombinant His fusion protein corresponding to human NCK1/NCK2.
100 uL
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human, Murine, Rat
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only