Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NEURL Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NEURL.
Supplier: Abnova Corporation PAB17798
Specifications
NEURL | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of NEURL. | |
In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
NEURL1/RNF67/h-neu/neu-1 | |
Rabbit | |
100 μg | |
Primary | |
This antibody detects endogenous levels of total NEURL protein. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
Western Blot (1:500-1:1000) ELISA (1:40000) The optimal working dilution should be determined by the end user. | |
NEURL | |
NEURL | |
A synthetic peptide corresponding to internal of human NEURL. | |
RUO | |
9148 | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction