Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Neurogenin-1, (WB dilu: 1:1000-1:10000), Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243787

Catalog No. NBP243787


Add to cart

Description

Description

Specifically detects Neurogenin-1 in Human samples, and it is validated for Western Blotting

Neurogenin-1 Polyclonal antibody specifically detects Neurogenin-1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Neurogenin-1
Unconjugated
PBS (pH 7.0) and 20% Glycerol with 0.01% Thimerosal
AKA, BHLHA6, bHLHa6NGN-1, Class A basic helix-loop-helix protein 6, Math4C, NeuroD3, NEUROD3neurogenic differentiation 3, Neurogenic basic-helix-loop-helix protein, Neurogenic differentiation factor 3, neurogenin 1, neurogenin-1, NGN, NGN1
Rabbit
IgG
0.1 ml
Neuroscience
Primary
4762
Western Blot
Western Blot 1:1000-1:10000
Ion Exchange Chromatography
NEUROG1
Recombinant protein encompassing a sequence within the center region of human Neurogenin 1. The exact sequence is proprietary.
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only