Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFKB1 Rabbit anti-Mouse, Rat, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of Nfkb1.

Manufacturer:  Abnova Corporation PAB11969

Catalog No. 89-115-478


Add to cart

Description

Description

Specifications

Specifications

NFKB1
Unconjugated
Western Blot (0.5 ug/mL) The optimal working dilution should be determined by the end user.
Nfkb1
NF-KB1|NF-kappaB|NF-kappaB1|p105|p50|p50/p105
Rabbit
25 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
This antibody detects a 50 KDa protein on mouse spleen tissue extract SDS-PAGE immunoblots, corresponding to the molecular weight of NFkB p50. The p105 precursor protein (105 KDa) may also be detected.
Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of Nfkb1.
In BBS, pH 8.2
Liquid
Nfkb1
A synthetic peptide corresponding to amino acids 4-21 of mouse Nfkb1.
RUO
Primary
18033
Murine, Rat
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only