Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOD2 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of NOD2.

Manufacturer:  Abnova Corporation PAB12097

Catalog No. 89-115-595


Add to cart

Description

Description

This gene is a member of the Nod1/Apaf-1 family and encodes a protein with two caspase recruitment (CARD) domains and six leucine-rich repeats (LRRs). The protein is primarily expressed in the peripheral blood leukocytes. It plays a role in the immune response to intracellular bacterial lipopolysaccharides (LPS) by recognizing the muramyl dipeptide (MDP) derived from them and activating the NFKB protein. Mutations in this gene have been associated with Crohn disease and Blau syndrome. [provided by RefSeq

Specifications

Specifications

NOD2
Unconjugated
Western Blot (0.5-2 ug/mL) The optimal working dilution should be determined by the end user.
NOD2
ACUG|BLAU|CARD15|CD|CLR16.3|IBD1|NLRC2|NOD2B|PSORAS1
Rabbit
100 uL
Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
This antibody is specific to NOD2 protein.
Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of NOD2.
In Tris-glycine, 150mM NaCl (0.05% sodium azide)
Liquid
NOD2
A synthetic peptide corresponding to amino acids 1000-1040 of human NOD2.
RUO
Primary
64127
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only