Learn More
NOS3 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Nos3.
Manufacturer:  Abnova Corporation PAB17901
Catalog No. 89-119-543
Add to cart
Description
Description
Specifications
Specifications
NOS3
Unconjugated
Western Blot (0.5-4 ug/mL) The optimal working dilution should be determined by the end user.
Nos3
eNos
Rabbit
100 ug
Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
This antibody recognizes 140 KDa eNOS of human, mouse, and rat origins.
Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of Nos3.
In PBS (30% glycerol, 0.5% BSA, 0.01% thimerosal)
Liquid
Nos3
A synthetic peptide corresponding to the residues surrounding amino acid 602 of rat Nos3.
RUO
Primary
24600
Human, Murine, Rat