Learn More
NOTUM Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NOTUM.
Manufacturer:  Abnova Corporation PAB16802
Catalog No. 89-118-621
Add to cart
Description
Description
Specifications
Specifications
NOTUM
Unconjugated
Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
NOTUM
NOTUM
A synthetic peptide corresponding to N-terminus 19 amino acids of human NOTUM.
RUO
Primary
147111
ELISA, Immunohistochemistry, Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of NOTUM.
In PBS (0.02% sodium azide)
Liquid
Rabbit
100 ug
Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human, Murine, Rat