Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NPBWR1 Rabbit anti-Human, Monkey, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NPBWR1.
Supplier: Abnova Corporation PAB26380
Specifications
NPBWR1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of NPBWR1. | |
In PBS (0.09% sodium azide) | |
GPR7/MGC129755 | |
Rabbit | |
Immunoaffinity chromatography | |
RUO | |
2831 | |
Human, Monkey | |
Liquid |
Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (11-22 ug/mL) The optimal working dilution should be determined by the end user. | |
NPBWR1 | |
NPBWR1 | |
A synthetic peptide corresponding to 17 amino acids at 2nd cytoplasmic domain of human NPBWR1. | |
50 μg | |
Primary | |
BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only