Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPHS1 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of NPHS1.

Manufacturer:  Abnova Corporation PAB14819

Catalog No. 89-117-286

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Nephrin is a kidney glomerular filtration barrier protein that is an essential component of the interpodocyte-spanning slit diaphragm. Mutations in the nephrin gene are associated with congenital nephrotic syndrome (NPHS1; MIM 256300).[supplied by OMIM

Specifications

Specifications

NPHS1
Lot specific
Rabbit polyclonal antibody raised against synthetic peptide of NPHS1.
In 10mM PBS, pH 7.4 (1% BSA, 0.09% sodium azide)
Liquid
NPHS1
A synthetic peptide corresponding to internal region of human NPHS1.
100 uL
Store at 4°C. Do not freeze.
Polyclonal
Molecular weight: 135 KDa. Epitope: Internal region.
Immunohistochemistry (PFA fixed)
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50) The optimal working dilution should be determined by the end user.
NPHS1
CNF|NPHN
Rabbit
Immunoaffinity purification
RUO
Primary
4868
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only