Learn More
NR1H3 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NR1H3.
Manufacturer:  Abnova Corporation PAB13685
Catalog No. 89-116-800
Add to cart
Description
Description
Specifications
Specifications
NR1H3
Unconjugated
Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
NR1H3
LXR-a|LXRA|RLD-1
Rabbit
IgG
100 ug
Store at -20°C on dry atmosphere. After reconstitution with 200μL of deionized water to a concentration of 500μg/mL
Polyclonal
Identical to the related rat and mouse sequence.
Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of NR1H3.
Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal)
Lyophilized
NR1H3
A synthetic peptide corresponding to amino acids at C-terminus of human NR1H3.
Affinity purification
RUO
Primary
10062
Human, Murine, Rat