Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NR1I3 Rabbit anti-Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Nr1i3.
Supplier: Abnova Corporation PAB5910
Specifications
NR1I3 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of Nr1i3. | |
In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide) | |
AA209988/AI551208/CAR/CAR-beta/Care2/ESTM32/MB67/MGC107281 | |
Rabbit | |
100 μg | |
Primary | |
Mouse, Rat | |
Liquid |
ELISA, Western Blot | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
Nr1i3 | |
Nr1i3 | |
A synthetic peptide corresponding to N-terminus of mouse Nr1i3. | |
RUO | |
12355 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only