Learn More
NRN1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NRN1.
Supplier: Abnova Corporation PAB12989
Description
This gene is expressed in postmitotic-differentiating neurons of the developmental nervous system and neuronal structures associated with plasticity in the adult. The expression of this gene can be induced by neural activity and neurotrophins. The encoded protein contains a consensus cleavage signal found in glycosylphoshatidylinositol (GPI)-anchored proteins. In vitro assays demonstrated that this protein promotes neurite outgrowth and arborization, suggesting its role in promoting neuritogenesis. [provided by RefSeq
Specifications
NRN1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of NRN1. | |
In PBS (0.02% sodium azide) | |
MGC44811/NRN/dJ380B8.2 | |
Rabbit | |
100 μg | |
Primary | |
Human, Mouse | |
Liquid |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
NRN1 | |
NRN1 | |
A synthetic peptide corresponding to internal region 20 amino acids of human NRN1. | |
RUO | |
51299 | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only