Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NRSN1, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of NRSN1.

Manufacturer:  Abnova Corporation PAB17800

Catalog No. 89-119-465


Add to cart

Description

Description

Specifications

Specifications

NRSN1
Unconjugated
Western Blot (1:500-1:1000) ELISA (1:40000) The optimal working dilution should be determined by the end user.
NRSN1
VMP|p24
Rabbit
100 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
This antibody detects endogenous levels of total NRSN1 protein
ELISA, Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of NRSN1.
In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol)
Liquid
NRSN1
A synthetic peptide corresponding to internal of human NRSN1.
RUO
Primary
140767
Human, Murine, Rat
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only