Learn More
Dengue Virus NS4B Rabbit anti-Virus, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242899
Catalog No. NBP242899
Add to cart
Description
Description
Specifically detects NS4B in Virus samples, and it is validated for Immunocytochemistry, Immunofluorescence, Western Blotting
Dengue Virus NS4B Polyclonal antibody specifically detects Dengue Virus NS4B in Virus samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.
Specifications
Specifications
Dengue Virus NS4B
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
1494449
Western Blot, Immunocytochemistry, Immunofluorescence
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:2000
Affinity Purified
Full length Japanese encephalitis virus JEV NS4b Recombinant protein
27 kDa
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Virus