Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dengue Virus NS4B Rabbit anti-Virus, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242897

Catalog No. NBP242897


Add to cart

Description

Description

Specifically detects NS4B in Virus samples, and it is validated for Western Blotting

Dengue Virus NS4B Polyclonal antibody specifically detects Dengue Virus NS4B in Virus samples. It is validated for Western Blot.
Specifications

Specifications

Dengue Virus NS4B
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
1494449
Western Blot
Western Blot 1:500-1:3000
Affinity Purified
The exact sequence is proprietary.
27 kDa
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Virus
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only