Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NTRK2 (phospho Y515), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic phosphopeptide of NTRK2.

Manufacturer:  Abnova Corporation PAB25893

Catalog No. 89-127-524


Add to cart

Description

Description

This gene encodes a member of the neurotrophic tyrosine receptor kinase (NTRK) family. This kinase is a membrane-bound receptor that, upon neurotrophin binding, phosphorylates itself and members of the MAPK pathway. Signalling through this kinase leads to cell differentiation. Mutations in this gene have been associated with obesity and mood disorders. Alternate transcriptional splice variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: P-Q-Y-Fp-G
Specifications

Specifications

NTRK2
1 mg/mL
Rabbit polyclonal antibody raised against synthetic phosphopeptide of NTRK2.
In PBS (without Mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide)
Liquid
NTRK2
Synthetic phosphopeptide corresponding to residues surrounding Y515 of human NTRK2.
100 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human, Murine, Rat
Immunofluorescence
Unconjugated
Immunofluorescence (1:100-1:200) The optimal working dilution should be determined by the end user.
NTRK2
GP145-TrkB|TRKB
Rabbit
Affinity chromatography
RUO
Primary
4915
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only