Learn More
NTSR1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NTSR1.
Supplier:  Abnova Corporation PAB26957
Catalog No. 89-128-603
Add to Cart
Description
Description
Neurotensin receptor 1 belongs to the large superfamily of G-protein coupled receptors. NTSR1 mediates the multiple functions of neurotensin, such as hypotension, hyperglycemia, hypothermia, antinociception, and regulation of intestinal motility and secretion. [provided by RefSeq
Specifications
Specifications
NTSR1
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of NTSR1.
In PBS, pH 7.2 (0.09% sodium azide)
NTR
Rabbit
Affinity purification
RUO
4923
Human, Mouse
Liquid
Immunofluorescence, Immunohistochemistry, Western Blot
Unconjugated
Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user.
NTSR1
NTSR1
A synthetic peptide corresponding to human NTSR1.
100 μL
Primary
NTSR1 polyclonal antibody detects endogenous levels of NTSR1 protein.
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Suggestions
Product Suggestions