Learn More
NUAK2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NUAK2.
Supplier:  Abnova Corporation PAB2716
Catalog No. 89-128-813
Add to Cart
Description
Description
Specifications
Specifications
NUAK2
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of NUAK2.
In PBS (0.09% sodium azide)
DKFZp434J037/DKFZp686F01113/FLJ90349/SNARK
Rabbit
Protein G purification
RUO
81788
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
ELISA, Western Blot
Unconjugated
Western Blot (1:1000) The optimal working dilution should be determined by the end user.
NUAK2
NUAK2
A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human NUAK2.
400 μL
Primary
Human
Liquid
Product Suggestions
Product Suggestions