Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUB1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of NUB1.

Manufacturer:  Abnova Corporation PAB27096

Catalog No. 89-128-747


Add to cart

Description

Description

NUB1 interacts with and negatively regulates NEDD8 (MIM 603171), a ubiquitin-like protein that covalently conjugates to cullin (see MIM 603134) family members.[supplied by OMIM]

Specifications

Specifications

NUB1
1 mg/mL
Rabbit polyclonal antibody raised against synthetic peptide of NUB1.
In PBS, pH 7.2 (0.05% sodium azide)
Liquid
NUB1
A synthetic peptide corresponding to human NUB1.
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
NUB1 polyclonal antibody detects endogenous levels of NUB1 protein.
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user.
NUB1
BS4|NUB1L|NYREN18
Rabbit
Antigen affinity purification
RUO
Primary
51667
Human, Murine
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only