Learn More
NUP107 Rabbit anti-Human, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NUP107.
Manufacturer:  Abnova Corporation PAB13393
Catalog No. 89-116-640
Add to cart
Description
Description
This gene encodes a member of the nucleoporin family. The protein is localized to the nuclear rim and is an essential component of the nuclear pore complex (NPC). All molecules entering or leaving the nucleus either diffuse through or are actively transported by the NPC. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized. [provided by RefSeq
Specifications
Specifications
NUP107
Unconjugated
Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
NUP107
NUP84
Rabbit
100 ug
Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human, Rat
Immunocytochemistry, Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of NUP107.
In PBS (0.02% sodium azide)
Liquid
NUP107
A synthetic peptide corresponding to C-terminus 15 amino acids of human NUP107.
RUO
Primary
57122