Learn More
OR10Z1, Rabbit, Polyclonal Antibody, Abnova
Rabbit polyclonal antibody raised against synthetic peptide of OR10Z1.
Manufacturer: Abnova Corporation PAB26989
Description
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms. [provided by RefSeq
Specifications
OR10Z1 | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of OR10Z1. | |
In PBS, pH 7.2 (0.05% sodium azide) | |
Liquid | |
OR10Z1 | |
A synthetic peptide corresponding to human OR10Z1. | |
100 uL | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Polyclonal | |
OR10Z1 polyclonal antibody detects endogenous levels of OR10Z1 protein. |
Western Blot | |
Unconjugated | |
Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
OR10Z1 | |
OR1-15 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
Primary | |
128368 | |
Human, Murine |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only