Learn More
OR4A16, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of OR4A16.
Supplier: Abnova Corporation PAB26968
Description
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms. [provided by RefSeq
Specifications
OR4A16 | |
Polyclonal | |
Unconjugated | |
Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
OR4A16 | |
OR4A16 | |
A synthetic peptide corresponding to human OR4A16. | |
100 μL | |
Primary | |
OR4A16 polyclonal antibody detects endogenous levels of OR4A16 protein. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of OR4A16. | |
In PBS, pH 7.2 (0.05% sodium azide) | |
OR11-117/OR4A16Q | |
Rabbit | |
Antigen affinity purification | |
RUO | |
81327 | |
Human, Mouse | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only