Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-OSBPL11, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236544

Catalog No. NBP236544


Add to cart

Description

Description

OSBPL11 Polyclonal antibody specifically detects OSBPL11 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

OSBPL11
1.0 mg/mL
Western Blot 1:2000 - 1: 10000, Immunoprecipitation 2 - 10 ug/mg
NP_073613.2
FLJ13012, FLJ13164, ORP11OSBP-related protein 11, ORP-11TCCCIA00292, OSBP12, oxysterol binding protein-like 11, oxysterol-binding protein-related protein 11
Rabbit
IgG
100 ul
Lipid and Metabolism
Primary
114885
Western Blot, Immunoprecipitation
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
OSBPL11
The immunogen recognized by the antibody maps to a region between residue 1 to 50 of human Oxysterol Binding Protein-Like 11 using the numbering given in entry NP_073613.2.
Immunogen affinity purified
RUO
Store at 4C. Do not freeze.
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit