Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-OVOL2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242907

Catalog No. NBP242907


Add to cart

Description

Description

Specifically detects OVOL2 in Human samples, and it is validated for Western Blotting

OVOL2 Polyclonal antibody specifically detects OVOL2 in Human, Mouse samples. It is validated for Western Blot.
Specifications

Specifications

OVOL2
Unconjugated
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
bA504H3.3, hOvo2, ovo-like 2 (Drosophila), transcription factor Ovo-like 2, Zinc finger protein 339EUROIMAGE566589, ZNF339
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
58495
Western Blot
Western Blot 1:500-1:3000
Affinity Purified
OVOL2
Recombinant protein encompassing a sequence within the center region of mouse mOvo2. The exact sequence is proprietary.
31 kDa
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only