Learn More
P2RY13 Rabbit anti-Human, Polyclonal Antibody, Abnova
Rabbit polyclonal antibody raised against synthetic peptide of P2RY13.
Manufacturer: Abnova Corporation PAB26505
Description
The product of this gene belongs to the family of G-protein coupled receptors. This family has several receptor subtypes with different pharmacological selectivity, which overlaps in some cases, for various adenosine and uridine nucleotides. This receptor is activated by ADP. [provided by RefSeq
Specifications
P2RY13 | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (11-22 ug/mL) The optimal working dilution should be determined by the end user. | |
P2RY13 | |
FKSG77|GPCR1|GPR86|GPR94|P2Y13|SP174 | |
Rabbit | |
Immunoaffinity chromatography | |
RUO | |
Primary | |
53829 | |
Human |
Immunohistochemistry (PFA fixed) | |
Rabbit polyclonal antibody raised against synthetic peptide of P2RY13. | |
In PBS (0.09% sodium azide) | |
Liquid | |
P2RY13 | |
A synthetic peptide corresponding to 15 amino acids at N-terminal extracellular domain of human P2RY13. | |
50 ug | |
Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
|
Polyclonal | |
BLAST analysis of the peptide immunogen showed no homology with other human proteins. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only