Learn More
PACS1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PACS1.
Supplier:  Abnova Corporation PAB5505
Catalog No. 89-131-883
Add to Cart
Description
Description
This gene encodes a protein with a putative role in the localization of trans-Golgi network (TGN) membrane proteins. Mouse and rat homologs have been identified and studies of the homologous rat protein indicate a role in directing TGN localization of furin by binding to the protease's phosphorylated cytosolic domain. In addition, the human protein plays a role in HIV-1 Nef-mediated downregulation of cell surface MHC-I molecules to the TGN, thereby enabling HIV-1 to escape immune surveillance. [provided by RefSeq
Specifications
Specifications
PACS1
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of PACS1.
In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide)
FLJ10209/KIAA1175
Rabbit
100 μg
Primary
Human, Mouse, Rat
Liquid
ELISA, Western Blot
Unconjugated
The optimal working dilution should be determined by the end user.
PACS1
PACS1
A synthetic peptide corresponding to C-terminus of human PACS1.
RUO
55690
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Recommendations
Recommendations