Learn More
PAPSS1, Rabbit, Polyclonal Antibody, Abnova
Rabbit polyclonal antibody raised against synthetic peptide of PAPSS1.
Manufacturer: Abnova Corporation PAB2534
Description
Three-prime-phosphoadenosine 5-prime-phosphosulfate (PAPS) is the sulfate donor cosubstrate for all sulfotransferase (SULT) enzymes (Xu et al., 2000 [PubMed 10679223]). SULTs catalyze the sulfate conjugation of many endogenous and exogenous compounds, including drugs and other xenobiotics. In humans, PAPS is synthesized from adenosine 5-prime triphosphate (ATP) and inorganic sulfate by 2 isoforms, PAPSS1 and PAPSS2 (MIM 603005).[supplied by OMIM
Specifications
PAPSS1 | |
Unconjugated | |
Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
PAPSS1 | |
ATPSK1|PAPSS|SK1 | |
Rabbit | |
Protein G purification | |
RUO | |
Primary | |
9061 |
ELISA, Western Blot | |
Rabbit polyclonal antibody raised against synthetic peptide of PAPSS1. | |
In PBS (0.09% sodium azide) | |
Liquid | |
PAPSS1 | |
A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human PAPSS1. | |
400 uL | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Polyclonal | |
Human |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only