Learn More
PARP1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova
Rabbit polyclonal antibody raised against synthetic peptide of PARP1.
Manufacturer: Abnova Corporation PAB27166
Description
This gene encodes a chromatin-associated enzyme, poly(ADP-ribosyl)transferase, which modifies various nuclear proteins by poly(ADP-ribosyl)ation. The modification is dependent on DNA and is involved in the regulation of various important cellular processes such as differentiation, proliferation, and tumor transformation and also in the regulation of the molecular events involved in the recovery of cell from DNA damage. In addition, this enzyme may be the site of mutation in Fanconi anemia, and may participate in the pathophysiology of type I diabetes. [provided by RefSeq
Specifications
PARP1 | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of PARP1. | |
In PBS, pH 7.2 (0.05% sodium azide) | |
Liquid | |
PARP1 | |
A synthetic peptide corresponding to human PARP1. | |
100 uL | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Polyclonal | |
PARP1 polyclonal antibody detects endogenous levels of PARP1 protein. |
Western Blot | |
Unconjugated | |
Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
PARP1 | |
ADPRT|ADPRT1|PARP|PARP-1|PPOL|pADPRT-1 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
Primary | |
142 | |
Human, Murine |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only