Learn More
PARP6 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PARP6.
Supplier:  Abnova Corporation PAB4873
Catalog No. 89-131-624
Add to Cart
Description
Description
Specifications
Specifications
PARP6
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of PARP6.
In PBS (0.09% sodium azide)
MGC131971
Rabbit
Ammonium sulfate precipitation
RUO
56965
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
ELISA, Western Blot
Unconjugated
ELISA (1:1000) Western Blot (1:10-50) The optimal working dilution should be determined by the end user.
PARP6
PARP6
A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human PARP6.
400 μL
Primary
Human
Liquid
Product Suggestions
Product Suggestions