Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-PCPE-2/PCOLCE2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242910

Catalog No. NBP242910


Add to cart

Description

Description

Specifically detects PCPE-2/PCOLCE2 in Human samples, and it is validated for Immunocytochemistry, Immunofluorescence

PCOLCE2 Polyclonal antibody specifically detects PCOLCE2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

PCPE-2/PCOLCE2
1mg/mL
Immunocytochemistry/Immunofluorescence 1:100-1:1000
Affinity Purified
Recombinant protein encompassing a sequence within the center region of human PCOLCE2. The exact sequence is proprietary.
46 kDa
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Human
Immunocytochemistry, Immunofluorescence
Unconjugated
PBS (pH 7.0) and 20% Glycerol with 0.01% Thimerosal
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
26577
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only