Learn More
PDCD6 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PDCD6.
Manufacturer:  Abnova Corporation PAB11731
Catalog No. 89-115-338
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
This gene encodes a calcium-binding protein belonging to the penta-EF-hand protein family. Calcium binding is important for homodimerization and for conformational changes required for binding to other protein partners. This gene product participates in T cell receptor-, Fas-, and glucocorticoid-induced programmed cell death. In mice deficient for this gene product, however, apoptosis was not blocked suggesting this gene product is functionally redundant. [provided by RefSeq
Specifications
Specifications
PDCD6
Lot specific
Rabbit polyclonal antibody raised against synthetic peptide of PDCD6.
In 10mM PBS, pH 7.4 (1% BSA, 0.09% sodium azide)
Liquid
PDCD6
A synthetic peptide corresponding to C-terminus of human PDCD6.
100 uL
Store at 4°C.
Polyclonal
Human
Immunohistochemistry (PFA fixed)
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (Formalin/PFA-fixed paraffin-embedded sections) (1:50) The optimal working dilution should be determined by the end user.
PDCD6
ALG-2|FLJ46208|MGC111017|MGC119050|MGC9123|PEF1B
Rabbit
Immunoaffinity purification
RUO
Primary
10016