Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pdgfb, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against recombinant Pdgfb.

Manufacturer:  Abnova Corporation PAB17984

Catalog No. 89-119-592


Add to cart

Description

Description

Specifications

Specifications

Pdgfb
Unconjugated
Western Blot (0.5-4 ug/mL) The optimal working dilution should be determined by the end user.
Pdgfb
PDGF-B|Sis
Rabbit
100 ug
Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Recombinant murine PDGF-BB can be used as a positive control.
Western Blot
Rabbit polyclonal antibody raised against recombinant Pdgfb.
In PBS pH 7.2 (30% glycerol, 0.01% thimerosal)
Liquid
Pdgfb
Recombinant protein corresponding to mouse Pdgfb.
RUO
Primary
18591
Mouse
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only