Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDGFRB (phospho Y771), Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic phosphopeptide of PDGFRB.

Manufacturer:  Abnova Corporation PAB1243

Catalog No. 89-115-803


Add to cart

Description

Description

This gene encodes a cell surface tyrosine kinase receptor for members of the platelet-derived growth factor family. These growth factors are mitogens for cells of mesenchymal origin. The identity of the growth factor bound to a receptor monomer determines whether the functional receptor is a homodimer or a heterodimer, composed of both platelet-derived growth factor receptor alpha and beta polypeptides. This gene is flanked on chromosome 5 by the genes for granulocyte-macrophage colony-stimulating factor and macrophage-colony stimulating factor receptor; all three genes may be implicated in the 5-q syndrome. A translocation between chromosomes 5 and 12, that fuses this gene to that of the translocation, ETV6, leukemia gene, results in chronic myeloproliferative disorder with eosinophilia. [provided by RefSeq

Specifications

Specifications

PDGFRB
Unconjugated
Western Blot (1:1800) The optimal working dilution should be determined by the end user.
PDGFRB
CD140B|JTK12|PDGF-R-beta|PDGFR|PDGFR1
Rabbit
100 uL
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human, Murine
Western Blot
Rabbit polyclonal antibody raised against synthetic phosphopeptide of PDGFRB.
In HEPES, 150mM NaCl (50% glycerol, 0.01% BSA, 0.03% sodium azide)
Liquid
PDGFRB
Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding Y771 of human PDGFRB.
RUO
Primary
5159
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only