Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pdzd3, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of Pdzd3.

Manufacturer:  Abnova Corporation PAB12076

Catalog No. 89-115-578


Add to cart

Description

Description

Specifications

Specifications

Pdzd3
Unconjugated
Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Pdzd3
NaPi-Cap2|Pdzk2
Rabbit
100 uL
Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
This antibody is specific to PDZK2 / IKEPP.
Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of Pdzd3.
In PBS (0.05% sodium azide)
Liquid
Pdzd3
A synthetic peptide corresponding to C-terminus of mouse Pdzd3.
RUO
Primary
170761
Human, Murine, Rat
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only