Learn More
PGK1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova
Rabbit polyclonal antibody raised against synthetic peptide of PGK1.
Manufacturer: Abnova Corporation PAB2618
Description
The protein encoded by this gene is a glycolytic enzyme that catalyzes the conversion of 1,3-diphosphoglycerate to 3-phosphoglycerate. The encoded protein may also act as a cofactor for polymerase alpha. This gene lies on the X-chromosome, while a related pseudogene also has been found on the X-chromosome and another on chromosome 19. [provided by RefSeq
Specifications
PGK1 | |
Unconjugated | |
Western Blot (1:1000) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
PGK1 | |
MGC117307|MGC142128|MGC8947|MIG10|PGKA | |
Rabbit | |
Protein G purification | |
RUO | |
Primary | |
5230 |
ELISA, Immunohistochemistry, Western Blot | |
Rabbit polyclonal antibody raised against synthetic peptide of PGK1. | |
In PBS (0.09% sodium azide) | |
Liquid | |
PGK1 | |
A synthetic peptide (conjugated with KLH) corresponding to internal region of human PGK1. | |
400 uL | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Polyclonal | |
Human, Murine |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only