Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-PGRMC1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236525

Catalog No. NBP236525


Add to cart

Description

Description

PGRMC1 Polyclonal antibody specifically detects PGRMC1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.
Specifications

Specifications

PGRMC1
1.0 mg/mL
Western Blot 1:1000 - 1:5000, Immunohistochemistry, Immunoprecipitation 2 - 10 ug/mg
NP_006658.1
HPR6.6PGRMC, membrane-associated progesterone receptor component 1, MPR, progesterone binding protein, progesterone receptor membrane component 1
Rabbit
IgG
100 ul
Cancer
Primary
10857
Western Blot, Immunohistochemistry, Immunoprecipitation
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
PGRMC1
The immunogen recognized by this antibody maps to a region between residue 145 to 195 of human Progesterone Receptor Membrane Component 1 using the numbering given in entry NP_006658.1.
Immunogen affinity purified
RUO
Store at 4C. Do not freeze.
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit