Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-PHLDB2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236546

Catalog No. NBP236546


Add to cart

Description

Description

PHLDB2 Polyclonal antibody specifically detects PHLDB2 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

PHLDB2
1.0 mg/mL
Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg
NP_001127910.1
pleckstrin homology-like domain, family B, member 2
Rabbit
IgG
100 ul
Store at 4C. Do not freeze.
Polyclonal
Human
Western Blot, Immunoprecipitation
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
PHLDB2
The immungoen recognized by the antibody maps to a region between residue 250 to 300 of human LL5beta using the numbering given in entry NP_001127910.1.
Immunogen affinity purified
RUO
Primary
90102
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit