Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PI4K2A Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP244158

Catalog No. NBP244158


Add to cart

Description

Description

Specifically detects PI4K2A in Human samples, and it is validated for ELISA, Western Blotting

PI4K2A Polyclonal antibody specifically detects PI4K2A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

PI4K2A
0.31 mg/ml
Western Blot 1 ug/ml, ELISA 1: 10000
Affinity Purified
PI4K2A
Anti-PI4K2A alpha affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide at the N-terminal of human PI4K2A protein.
Immunogen affinity purified
RUO
Primary
55361
Western Blot, ELISA
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 50% (v/v) Glycerol with No Preservative
DKFZp761G1923, EC 2.7.1.67, phosphatidylinositol 4-kinase type 2 alpha, phosphatidylinositol 4-kinase type 2-alpha, phosphatidylinositol 4-kinase type II (PI4KII), Phosphatidylinositol 4-kinase type II-alpha, PI4KII, PIK42A, RP11-548K23.6
Rabbit
IgG
0.1 mg
Store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only