Learn More
PICK1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PICK1.
Supplier: Abnova Corporation PAB4632
Description
The protein encoded by this gene contains a PDZ domain, through which it interacts with protein kinase C, alpha (PRKCA). This protein may function as an adaptor that binds to and organizes the subcellular localization of a variety of membrane proteins. It has been shown to interact with multiple glutamate receptor subtypes, monoamine plasma membrane transporters, as well as nonvoltage gated sodium channels, and may target PRKCA to these membrane proteins and thus regulate their distribution and function. This protein has also been found to act as an anchoring protein that specifically targets PRKCA to mitochondria in a ligand-specific manner. Three transcript variants encoding the same protein have been found for this gene. [provided by RefSeq
Specifications
PICK1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of PICK1. | |
In PBS (0.09% sodium azide) | |
MGC15204/PICK/PRKCABP | |
Rabbit | |
Protein G purification | |
RUO | |
9463 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
ELISA (1:1000) Western Blot (1:100-500) The optimal working dilution should be determined by the end user. | |
PICK1 | |
PICK1 | |
A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human PICK1. | |
400 μL | |
Primary | |
Human, Mouse | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only