Learn More
PIGY Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PIGY.
Supplier: Abnova Corporation PAB16600
Description
This gene encodes two proteins, one of which is part of the GPI-N-acetylglucosaminyltransferase (GIP-GnT) complex which initiates the biosynthesis of glycosylphosphatidylinositol (GPI). GPI is synthesized in the endoplasmic reticulum and serves as an anchor for many surface proteins. Proteins containing GPI anchors can have an important role in cell-cell interactions. Two open reading frames have been found in the single transcript that has been identified for this gene. The downstream open reading frame encodes the GPI-GnT complex protein while the upstream open reading frame encodes a protein with unknown function. [provided by RefSeq
Specifications
PIGY | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of PIGY. | |
In PBS (0.02% sodium azide) | |
MGC14156/PIG-Y | |
Rabbit | |
100 μg | |
Primary | |
Human, Mouse | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
Unconjugated | |
Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
PIGY | |
PIGY | |
A synthetic peptide corresponding to internal region 16 amino acids of human PIGY. | |
RUO | |
84992 | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only